Assay Details
| Gene Symbol: C1RL-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026947.1 | 8 | 2524 | 105 | ||
| GenBank mRNA | AL833364.1 | - | 8 | 2524 | 105 | |
| Gene Symbol: RBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096947.1 | - | 4 | 105 | ||
More Information
Set Membership: |
|

C1RL-AS1
Genomic Map