Assay Details
| Gene Symbol: UBQLN2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013444.3 | NP_038472.2 | 1 | 2378 | 114 | |
| GenBank mRNA | AF189009.1 | - | 1 | 2332 | 114 | |
| AK001029.1 | - | 2 | 945 | 114 | ||
| AK297252.1 | - | 2 | 2025 | 114 | ||
| AL442081.1 | - | 1 | 1991 | 114 | ||
| BC069237.1 | - | 2 | 2249 | 4543266 | 114 | |
More Information
Set Membership: |
|

UBQLN2
Genomic Map