Assay Details
| Gene Symbol: IGBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001551.2 | NP_001542.1 | 2 | 182 | 63 | |
| XM_017029489.1 | XP_016884978.1 | 1 | 383 | 63 | ||
| XM_017029490.1 | XP_016884979.1 | 1 | 383 | 63 | ||
| GenBank mRNA | AK054596.1 | - | 1 | 383 | 63 | |
| AK315660.1 | - | 1 | 300 | 63 | ||
| BC004137.2 | - | 2 | 182 | 2962967 | 63 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the AK054596.1, AK315660.1, BC004137.2, NM_001551.2, XM_017029489.1, XM_017029490.1 transcripts.
Set Membership: |
|

IGBP1
Genomic Map