Assay Details
| Gene Symbol: PPEF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_006239.2 | NP_006230.2 | 8 | 1057 | 101 | |
| XM_011532039.2 | XP_011530341.1 | 7 | 1290 | 101 | ||
| GenBank mRNA | AF023456.1 | - | 8 | 1067 | 101 | |
| AF023457.1 | - | 7 | 700 | 101 | ||
| BX648754.1 | - | 8 | 977 | 101 | ||
More Information
Set Membership: |
|

PPEF2
Genomic Map