Assay Details
| Gene Symbol: LOC103171574 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120379.1 | 2 - 3 | 251 | 103 | ||
| GenBank mRNA | AK311032.1 | - | 2 - 3 | 152 | 103 | |
| Gene Symbol: LOC102724034 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120378.1 | 2 - 3 | 152 | 103 | ||
| GenBank mRNA | DC422276.1 | - | 2 - 3 | 152 | 103 | |
More Information
Set Membership: |
|

LOC103171574
Genomic Map