Assay Details
| Gene Symbol: EWSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096787.1 | - | 3 | 1213 | 65 | |
| AL833489.1 | - | 6 | 3616 | 65 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EWSR1| Gene Symbol: EWSR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK096787.1 | - | 3 | 1213 | 65 | |
| AL833489.1 | - | 6 | 3616 | 65 | ||
Set Membership: |
|