Assay Details
| Gene Symbol: NEFH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_021076.3 | NP_066554.2 | 1 | 797 | 84 | |
| XM_011530200.2 | XP_011528502.1 | 1 | 763 | 84 | ||
| GenBank mRNA | AB020652.2 | - | 3 | 991 | 84 | |
| AF203032.1 | - | 1 | 726 | 84 | ||
| AK302660.1 | - | 1 | 797 | 84 | ||
| BC073969.1 | - | 1 | 759 | 6137962 | 84 | |
More Information
Set Membership: |
|

NEFH
Genomic Map