Assay Details
| Gene Symbol: ATP6V1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001690.3 | NP_001681.2 | 15 | 2970 | 104 | |
| GenBank mRNA | AF113129.1 | - | 15 | 2900 | 104 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ATP6V1A| Gene Symbol: ATP6V1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001690.3 | NP_001681.2 | 15 | 2970 | 104 | |
| GenBank mRNA | AF113129.1 | - | 15 | 2900 | 104 | |
Set Membership: |
|