Assay Details
| Gene Symbol: PLCXD3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001005473.2 | NP_001005473.1 | 3 | 3117 | 106 | |
| GenBank mRNA | BX648329.1 | - | 4 | 2928 | 106 | |
More Information
Set Membership: |
|

PLCXD3
Genomic Map