Assay Details
| Gene Symbol: EFR3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001323554.1 | NP_001310483.1 | 1 | 1083 | 101 | |
| NM_001323555.1 | NP_001310484.1 | 1 | 1083 | 101 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001323554.1, NM_001323555.1 transcripts.
Set Membership: |
|

EFR3A
Genomic Map