Assay Details
| Gene Symbol: PSAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042465.2 | NP_001035930.1 | 15 | 2256 | 109 | |
| NM_001042466.2 | NP_001035931.1 | 15 | 2253 | 109 | ||
| NM_002778.3 | NP_002769.1 | 14 | 2247 | 109 | ||
| GenBank mRNA | AK129790.1 | - | 15 | 2130 | 109 | |
| BC001503.2 | - | 14 | 2098 | 3508263 | 109 | |
| BC004275.1 | - | 14 | 2106 | 3615413 | 109 | |
| BC007612.1 | - | 14 | 2111 | 3351606 | 109 | |
| BC091483.1 | - | 9 | 1293 | 109 | ||
| D00422.1 | - | 15 | 2110 | 109 | ||
| J03015.1 | - | 14 | 2100 | 109 | ||
| J03077.1 | - | 14 | 2138 | 109 | ||
| M32221.1 | - | 14 | 2100 | 109 | ||
| M60255.1 | - | 15 | 2102 | 109 | ||
| M60257.1 | - | 14 | 2093 | 109 | ||
| M60258.1 | - | 15 | 2099 | 109 | ||
| M81355.1 | - | 15 | 2110 | 109 | ||
More Information
Set Membership: |
|

PSAP
Genomic Map