Assay Details
| Gene Symbol: ZNF135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164529.1 | NP_001158001.1 | 4 | 500 | 114 | |
| NM_001164530.1 | NP_001158002.1 | 4 | 859 | 114 | ||
| NM_001289401.1 | NP_001276330.1 | 5 | 926 | 114 | ||
| NM_001289402.1 | NP_001276331.1 | 5 | 1289 | 114 | ||
| NM_003436.3 | NP_003427.3 | 5 | 957 | 114 | ||
| NM_007134.1 | NP_009065.1 | 4 | 895 | 114 | ||
| XM_005259211.1 | XP_005259268.1 | 4 | 859 | 114 | ||
| XM_006723362.3 | XP_006723425.1 | 5 | 965 | 114 | ||
| XM_006723363.3 | XP_006723426.1 | 5 | 1329 | 114 | ||
| XM_017027240.1 | XP_016882729.1 | 4 | 1282 | 114 | ||
| XM_017027241.1 | XP_016882730.1 | 5 | 1284 | 114 | ||
| XM_017027242.1 | XP_016882731.1 | 4 | 724 | 114 | ||
| GenBank mRNA | AK094202.1 | - | 5 | 1248 | 114 | |
| AK098011.1 | - | 4 | 859 | 114 | ||
| AK295110.1 | - | 5 | 927 | 114 | ||
| AK297226.1 | - | 4 | 690 | 114 | ||
| AL833637.1 | - | 4 | 458 | 114 | ||
| BC046434.1 | - | 5 | 921 | 4824328 | 114 | |
| BX647775.1 | - | 4 | 863 | 114 | ||
| U09413.1 | - | 1 | 328 | 114 | ||
More Information
Set Membership: |
|

ZNF135
Genomic Map