Assay Details
| Gene Symbol: LOC143666 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_026967.1 | 1 | 1226 | 73 | ||
| GenBank mRNA | AK057794.1 | - | 2 | 809 | 73 | |
| AK123970.1 | - | 1 | 1225 | 73 | ||
| BC018884.2 | - | 1 | 203 | 73 | ||
More Information
Set Membership: |
|

LOC143666
Genomic Map