Assay Details
| Gene Symbol: MIR9-3HG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_015411.1 | 7 | 3082 | 108 | ||
| NR_133001.1 | 7 | 3662 | 108 | |||
| NR_133002.1 | 6 | 2963 | 108 | |||
| NR_133003.1 | 7 | 2997 | 108 | |||
| GenBank mRNA | AK054710.1 | - | 1 | 698 | 108 | |
| AK096359.1 | - | 1 | 1197 | 108 | ||
More Information
Set Membership: |
|

MIR9-3HG
Genomic Map