Assay Details
| Gene Symbol: WASH1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011517662.2 | XP_011515964.1 | 1 | 935 | 129 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011517662.2 transcripts.
Set Membership: |
|

WASH1
Genomic Map