Assay Details
| Gene Symbol: CARHSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042476.2 | NP_001035941.1 | 4 | 2432 | 104 | |
| NM_001278260.1 | NP_001265189.1 | 5 | 2240 | 104 | ||
| NM_001278261.1 | NP_001265190.1 | 5 | 2379 | 104 | ||
| NM_001278262.1 | NP_001265191.1 | 5 | 2338 | 104 | ||
| NM_001278263.1 | NP_001265192.1 | 4 | 2114 | 104 | ||
| NM_001278264.1 | NP_001265193.1 | 4 | 2256 | 104 | ||
| NM_001278265.1 | NP_001265194.1 | 4 | 2248 | 104 | ||
| NM_001278266.1 | NP_001265195.1 | 4 | 2224 | 104 | ||
| NM_014316.3 | NP_055131.2 | 4 | 2126 | 104 | ||
| XM_005255228.4 | XP_005255285.3 | 4 | 2290 | 104 | ||
| XM_005255229.4 | XP_005255286.1 | 4 | 2102 | 104 | ||
| XM_011522444.2 | XP_011520746.1 | 4 | 2101 | 104 | ||
| GenBank mRNA | BC108283.1 | - | 4 | 2113 | 4430943 | 104 |
| BQ004670.1 | - | 1 | 104 | |||
More Information
Set Membership: |
|

CARHSP1
Genomic Map