Assay Details
| Gene Symbol: ADAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001324444.1 | NP_001311373.1 | 11 | 2399 | 65 | |
| NM_001324445.1 | NP_001311374.1 | 10 | 3114 | 65 | ||
| NM_001324446.1 | NP_001311375.1 | 11 | 3132 | 65 | ||
| NM_001324448.1 | NP_001311377.1 | 9 | 2095 | 65 | ||
| NM_001324449.1 | NP_001311378.1 | 8 | 2868 | 65 | ||
| NM_001324450.1 | NP_001311379.1 | 9 | 2153 | 65 | ||
| NM_001324451.1 | NP_001311380.1 | 8 | 2926 | 65 | ||
| NM_001324452.1 | NP_001311381.1 | 9 | 3059 | 65 | ||
| NM_001324453.1 | NP_001311382.1 | 7 | 2813 | 65 | ||
| NM_012091.4 | NP_036223.2 | 11 | 2341 | 65 | ||
| GenBank mRNA | AK001687.1 | - | 11 | 2332 | 65 | |
More Information
Set Membership: |
|

ADAT1
Genomic Map