Assay Details
Gene Symbol: SMCR5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_024007.1 | 1 | 482 | 104 | ||
GenBank mRNA | AF467442.1 | - | 1 | 482 | 104 |

More Information
Additional Information:
This assay is in the 5'UTR region of the AF467442.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
