Assay Details
| Gene Symbol: LYST | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000081.3 | NP_000072.2 | 12 | 4591 | 107 | |
| NM_001301365.1 | NP_001288294.1 | 12 | 5028 | 107 | ||
| XM_011544031.1 | XP_011542333.1 | 12 | 4591 | 107 | ||
| XM_011544032.1 | XP_011542334.1 | 12 | 4994 | 107 | ||
| XM_011544033.2 | XP_011542335.1 | 12 | 4741 | 107 | ||
| XM_011544034.1 | XP_011542336.1 | 12 | 4591 | 107 | ||
| XM_011544035.2 | XP_011542337.1 | 12 | 4591 | 107 | ||
| XM_011544036.2 | XP_011542338.1 | 8 | 2245 | 107 | ||
| XM_011544037.2 | XP_011542339.1 | 12 | 4591 | 107 | ||
| XM_011544039.2 | XP_011542341.1 | 12 | 4591 | 107 | ||
| XM_011544040.2 | XP_011542342.1 | 12 | 4591 | 107 | ||
| XM_017000150.1 | XP_016855639.1 | 12 | 4591 | 107 | ||
| XM_017000151.1 | XP_016855640.1 | 12 | 4591 | 107 | ||
| GenBank mRNA | AK307457.1 | - | 12 | 4994 | 107 | |
| U67615.1 | - | 12 | 4598 | 107 | ||
| U70064.1 | - | 12 | 4961 | 107 | ||
| U84744.1 | - | 12 | 4961 | 107 | ||
More Information
Set Membership: |
|

LYST
Genomic Map