Assay Details
| Gene Symbol: TRAF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004295.3 | NP_004286.2 | 7 | 1799 | 93 | |
| XM_011525504.2 | XP_011523806.1 | 7 | 1822 | 93 | ||
| XM_011525505.2 | XP_011523807.1 | 7 | 3617 | 93 | ||
| XM_011525506.2 | XP_011523808.1 | 7 | 3594 | 93 | ||
| XM_011525507.2 | XP_011523809.1 | 7 | 2168 | 93 | ||
| XM_011525508.2 | XP_011523810.1 | 7 | 1885 | 93 | ||
| GenBank mRNA | AK300804.1 | - | 8 | 1275 | 93 | |
| BC001769.1 | - | 7 | 1783 | 3354343 | 93 | |
| BC026726.1 | - | 4 | 1307 | 93 | ||
| BC047358.1 | - | 4 | 1307 | 93 | ||
| X80200.1 | - | 7 | 1776 | 93 | ||
More Information
Set Membership: |
|

TRAF4
Genomic Map