Assay Details
Gene Symbol: TMEM200C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011525729.2 | XP_011524031.1 | 3 | 5316 | 108 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011525729.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
