Assay Details
| Gene Symbol: GUCY1A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_000855.2 | NP_000846.1 | 8 | 9323 | 125 | |
| NM_001256424.1 | NP_001243353.1 | 9 | 9416 | 125 | ||
More Information
Set Membership: |
|

GUCY1A2
Genomic Map