Assay Details
| Gene Symbol: ADM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101340.1 | NP_001094810.1 | 2 | 1577 | 92 | |
| GenBank mRNA | AL359598.1 | - | 2 | 1078 | 92 | |
| BC032764.1 | - | 2 | 1609 | 92 | ||
| BC112359.1 | - | 2 | 1298 | 92 | ||
More Information
Set Membership: |
|

ADM5
Genomic Map