Assay Details
Gene Symbol: EMC10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_175063.5 | NP_778233.4 | 8 | 1916 | 72 | |
NM_206538.3 | NP_996261.1 | 7 | 1829 | 72 | ||
GenBank mRNA | AL512761.1 | - | 4 | 1432 | 72 | |
AY194293.2 | - | 12 | 2874 | 72 | ||
AY358710.1 | - | 8 | 966 | 72 | ||
AY761095.1 | - | 8 | 1894 | 72 | ||
AY761097.1 | - | 7 | 1807 | 72 | ||
BC032948.2 | - | 7 | 1768 | 4794992 | 72 | |
BC035001.2 | - | 7 | 1766 | 4796476 | 72 | |
BC062607.1 | - | 8 | 2058 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
