Assay Details
Gene Symbol: IPO13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_014652.3 | NP_055467.3 | 20 | 3912 | 61 | |
GenBank mRNA | AB018267.1 | - | 20 | 3779 | 61 | |
AK023441.1 | - | 14 | 1635 | 61 | ||
BX116797.1 | - | 3 | 540 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
