Assay Details
Gene Symbol: SCN2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017004659.1 | XP_016860148.1 | 1 | 222 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017004659.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
