Assay Details
| Gene Symbol: PDP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161780.1 | NP_001155252.1 | 2 - 3 | 184 | 66 | |
| GenBank mRNA | BC064978.1 | - | 2 - 3 | 168 | 66 | |
| DA507397.1 | - | 2 - 3 | 184 | 66 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001161780.1 transcripts.
Set Membership: |
|

PDP1
Genomic Map