Assay Details
Gene Symbol: BLOC1S6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001311255.1 | NP_001298184.1 | 4 - 5 | 559 | 95 | |
NM_001311256.1 | NP_001298185.1 | 3 - 4 | 471 | 95 | ||
NM_012388.3 | NP_036520.1 | 4 - 5 | 725 | 95 | ||
NR_132350.1 | 5 - 6 | 733 | 95 | |||
NR_132351.1 | 4 - 5 | 721 | 95 | |||
NR_132352.1 | 3 - 4 | 637 | 95 | |||
NR_132353.1 | 3 - 4 | 583 | 95 | |||
NR_132354.1 | 3 - 4 | 579 | 95 | |||
NR_132355.1 | 2 - 3 | 495 | 95 | |||
NR_132356.1 | 4 - 5 | 427 | 95 | |||
NR_132358.1 | 2 - 3 | 329 | 95 | |||
NR_132359.1 | 2 - 3 | 197 | 95 | |||
GenBank mRNA | AF080470.1 | - | 4 - 5 | 629 | 95 | |
AK057545.1 | - | 4 - 5 | 465 | 95 | ||
AK128626.1 | - | 5 - 6 | 702 | 95 | ||
AK315581.1 | - | 4 - 5 | 443 | 95 | ||
AL596952.1 | - | 4 - 5 | 498 | 95 | ||
BC004819.1 | - | 4 - 5 | 446 | 3450397 | 95 | |
DA226593.1 | - | 2 - 3 | 267 | 95 | ||
DA230439.1 | - | 2 - 3 | 330 | 95 | ||
DA233328.1 | - | 2 - 3 | 186 | 95 | ||
DA247079.1 | - | 3 - 4 | 357 | 95 | ||
DA523523.1 | - | 3 - 4 | 285 | 95 | ||
DA798595.1 | - | 3 - 4 | 351 | 95 | ||
DB212570.1 | - | 3 - 4 | 470 | 95 | ||
DB287075.1 | - | 2 - 3 | 268 | 95 | ||
DB289156.1 | - | 4 - 5 | 370 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
