Assay Details
| Gene Symbol: SEC22B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004892.5 | NP_004883.3 | 2 - 3 | 322 | 89 | |
| XM_017002856.1 | XP_016858345.1 | 2 - 3 | 335 | 89 | ||
| XM_017002857.1 | XP_016858346.1 | 2 - 3 | 764 | 89 | ||
| GenBank mRNA | AF047442.1 | - | 2 - 3 | 246 | 89 | |
| AK289875.1 | - | 2 - 3 | 316 | 89 | ||
| BC001364.2 | - | 2 - 3 | 301 | 3051087 | 89 | |
| CD619408.1 | - | 2 - 3 | 352 | 89 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017002857.1 transcripts.
Set Membership: |
|

SEC22B
Genomic Map