Assay Details
| Gene Symbol: GLRX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199868.1 | NP_001186797.1 | 11 - 12 | 1086 | 69 | |
| XM_017015474.1 | XP_016870963.1 | 12 - 13 | 1141 | 69 | ||
| GenBank mRNA | AJ010841.1 | - | 11 - 12 | 1044 | 69 | |
| AK022131.1 | - | 11 - 12 | 1060 | 69 | ||
| BI256922.1 | - | 6 - 7 | 427 | 69 | ||
More Information
Set Membership: |
|

GLRX3
Genomic Map