Assay Details
| Gene Symbol: CDKL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001009565.1 | NP_001009565.1 | 3 - 4 | 369 | 120 | |
| XM_011532815.2 | XP_011531117.1 | 4 - 5 | 1785 | 120 | ||
| XM_011532816.2 | XP_011531118.1 | 4 - 5 | 1785 | 120 | ||
| XM_011532817.2 | XP_011531119.1 | 4 - 5 | 1785 | 120 | ||
| XM_011532819.2 | XP_011531121.1 | 4 - 5 | 1785 | 120 | ||
| XM_017003981.1 | XP_016859470.1 | 4 - 5 | 1137 | 120 | ||
| XM_017003982.1 | XP_016859471.1 | 4 - 5 | 613 | 120 | ||
| XM_017003983.1 | XP_016859472.1 | 4 - 5 | 1785 | 120 | ||
| XM_017003984.1 | XP_016859473.1 | 4 - 5 | 1785 | 120 | ||
| XM_017003985.1 | XP_016859474.1 | 4 - 5 | 1785 | 120 | ||
| GenBank mRNA | AY845084.1 | - | 3 - 4 | 369 | 120 | |
| AY847283.1 | - | 3 - 4 | 369 | 120 | ||
| BC144460.1 | - | 3 - 4 | 433 | 120 | ||
More Information
Set Membership: |
|

CDKL4
Genomic Map