Assay Details
| Gene Symbol: LOC100507006 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120420.1 | 4 - 5 | 666 | 133 | ||
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Clariom D Assay | PSR0200153823.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153823.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153823.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153823.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153823.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153823.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153823.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153823.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153823.hg.1 | TC0200007792.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153814.hg.1 | TC0200007792.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153819.hg.1 | TC0200007792.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000438115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000438115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000438115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000438115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153813.hg.1 | TC0200007792.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200153808.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153808.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153808.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153808.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153808.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153808.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153808.hg.1 | TC0200007792.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153818.hg.1 | TC0200007792.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200153810.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153810.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153810.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153810.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153810.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153810.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153810.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153810.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153810.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153810.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153810.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153810.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153810.hg.1 | TC0200007792.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200153824.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153824.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153824.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153824.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153824.hg.1 | TC0200007792.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200153827.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153827.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153827.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200153827.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200153827.hg.1 | TC0200007792.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000413030.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000426365.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000436950.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000438115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000438115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000438115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000438115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000438115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000438115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000438115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000438115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000438115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000438115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000438115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000438115.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000449678.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | ENST00000451350.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | NR_120420.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200007792.hg.1 | NR_120420.1 | Refseq |

LOC100507006
Genomic Map