Assay Details
| Gene Symbol: LINCR-0002 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_120606.1 | 6 - 7 | 733 | 72 | ||
| GenBank mRNA | BX365943.2 | - | 3 - 4 | 451 | 72 | |
More Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Clariom D Assay | PSR0300171791.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171791.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171791.hg.1 | TC0300009868.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300171814.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171814.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171814.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171814.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171814.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171814.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171814.hg.1 | TC0300009868.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300171787.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171787.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171787.hg.1 | TC0300009868.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300171815.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171815.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171815.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171815.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171815.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171815.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171815.hg.1 | TC0300009868.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300171808.hg.1 | ENST00000438488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171808.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171808.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171808.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171808.hg.1 | ENST00000438488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171808.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171808.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171808.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171808.hg.1 | TC0300009868.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300171792.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171792.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171792.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171792.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171792.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171792.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171792.hg.1 | TC0300009868.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300171795.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171795.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171795.hg.1 | TC0300009868.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300171804.hg.1 | ENST00000438488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171804.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171804.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171804.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171804.hg.1 | ENST00000438488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171804.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171804.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171804.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171804.hg.1 | TC0300009868.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000438488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000438488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000438488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000438488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000438488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300009868.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171813.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171813.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171813.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171813.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171813.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171813.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171813.hg.1 | TC0300009868.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300171785.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171785.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171785.hg.1 | TC0300009868.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300171812.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171812.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171812.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171812.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171812.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171812.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171812.hg.1 | TC0300009868.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0300171809.hg.1 | ENST00000438488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171809.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171809.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171809.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171809.hg.1 | ENST00000438488.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171809.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171809.hg.1 | ENST00000439804.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0300171809.hg.1 | NR_120606.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0300171809.hg.1 | TC0300009868.hg.1 | Clariom D TC |

LINCR-0002
Genomic Map