Assay Details
| Gene Symbol: ST6GAL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011513085.1 | XP_011511387.1 | 2 - 3 | 575 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011513085.1 transcripts.
Set Membership: |
|

ST6GAL1
Genomic Map