Assay Details
Gene Symbol: CLDN6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021195.4 | NP_067018.2 | 2 | 122 | ||
GenBank mRNA | AF125306.1 | - | 1 | 122 | ||
AK075329.1 | - | 2 | 122 | |||
AY358480.1 | - | 1 | 122 | |||
BC008934.2 | - | 1 | 2963008 | 122 | ||
BT007399.1 | - | 1 | 122 | |||
BX444798.2 | - | 2 | 479 | 122 | ||
CN420342.1 | - | 1 | 122 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
