Assay Details
| Gene Symbol: LOC100996724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017003064.1 | XP_016858553.1 | 46 | 9528 | 128 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100996724| Gene Symbol: LOC100996724 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017003064.1 | XP_016858553.1 | 46 | 9528 | 128 | |
Set Membership: |
|