Assay Details
Gene Symbol: LOC100506022 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NR_104622.2 | 2 | 1666 | 67 | ||
NR_104623.2 | 2 | 1666 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Clariom D Assay | PSR0100194652.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194652.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194652.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194652.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194652.hg.1 | TC0100012801.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100194648.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194648.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194648.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194648.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194648.hg.1 | TC0100012801.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100194645.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194645.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194645.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194645.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194645.hg.1 | TC0100012801.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100194654.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194654.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194654.hg.1 | TC0100012801.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100194650.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194650.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194650.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194650.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194650.hg.1 | TC0100012801.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100194651.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194651.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194651.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194651.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194651.hg.1 | TC0100012801.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100194656.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194656.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194656.hg.1 | TC0100012801.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100194646.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194646.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194646.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194646.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194646.hg.1 | TC0100012801.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100194649.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194649.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194649.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194649.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194649.hg.1 | TC0100012801.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | TC0100012801.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194647.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194647.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194647.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194647.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194647.hg.1 | TC0100012801.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0100194653.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194653.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194653.hg.1 | NR_104622.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194653.hg.1 | NR_104623.2 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100194653.hg.1 | TC0100012801.hg.1 | Clariom D TC |
