Assay Details
| Gene Symbol: LINC00083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017003086.1 | XP_016858575.1 | 5 - 6 | 694 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00083| Gene Symbol: LINC00083 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017003086.1 | XP_016858575.1 | 5 - 6 | 694 | 75 | |
Set Membership: |
|