Assay Details
Gene Symbol: C2orf88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005246905.1 | XP_005246962.1 | 2 - 3 | 269 | 84 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005246905.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
