Assay Details
| Gene Symbol: SPAG16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024532.4 | NP_078808.3 | 14 - 15 | 1700 | 107 | |
| NR_047659.1 | 16 - 17 | 1875 | 107 | |||
| NR_047660.1 | 13 - 14 | 1581 | 107 | |||
| XM_011511814.1 | XP_011510116.1 | 13 - 14 | 1748 | 107 | ||
| XM_011511815.1 | XP_011510117.1 | 14 - 15 | 1651 | 107 | ||
| XM_011511816.2 | XP_011510118.1 | 14 - 15 | 1651 | 107 | ||
| XM_011511817.1 | XP_011510119.1 | 14 - 15 | 1651 | 107 | ||
| XM_011511818.1 | XP_011510120.1 | 14 - 15 | 1651 | 107 | ||
| XM_011511819.2 | XP_011510121.1 | 13 - 14 | 2399 | 107 | ||
| XM_011511820.1 | XP_011510122.1 | 13 - 14 | 1465 | 107 | ||
| XM_011511821.2 | XP_011510123.1 | 13 - 14 | 2525 | 107 | ||
| XM_011511822.1 | XP_011510124.1 | 12 - 13 | 1381 | 107 | ||
| XM_011511823.2 | XP_011510125.1 | 12 - 13 | 2303 | 107 | ||
| XM_011511835.2 | XP_011510137.1 | 12 - 13 | 1633 | 107 | ||
| XM_017004896.1 | XP_016860385.1 | 15 - 16 | 1780 | 107 | ||
| XM_017004897.1 | XP_016860386.1 | 12 - 13 | 4397 | 107 | ||
| XM_017004898.1 | XP_016860387.1 | 9 - 10 | 1129 | 107 | ||
| XM_017004900.1 | XP_016860389.1 | 13 - 14 | 1532 | 107 | ||
| XM_017004901.1 | XP_016860390.1 | 13 - 14 | 1514 | 107 | ||
| XM_017004902.1 | XP_016860391.1 | 7 - 8 | 850 | 107 | ||
| GenBank mRNA | AF310672.1 | - | 14 - 15 | 1637 | 107 | |
| AF490390.1 | - | 14 - 15 | 1630 | 107 | ||
| AK302351.1 | - | 13 - 14 | 1523 | 107 | ||
| BC025379.1 | - | 16 - 17 | 1880 | 4820489 | 107 | |
| CR749477.1 | - | 12 - 13 | 1386 | 107 | ||
More Information
Set Membership: |
|

SPAG16
Genomic Map