Assay Details
| Gene Symbol: XPC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_004628.4 | NP_004619.3 | 9 | 1409 | 116 | |
| NR_027299.1 | 8 | 1285 | 116 | |||
| XM_011534093.2 | XP_011532395.1 | 9 | 1366 | 116 | ||
| GenBank mRNA | AK222844.1 | - | 8 | 1102 | 116 | |
| AK289761.1 | - | 9 | 116 | |||
| AK295711.1 | - | 9 | 1227 | 116 | ||
| AK311039.1 | - | 8 | 1214 | 116 | ||
| BC016620.1 | - | 9 | 1320 | 4509957 | 116 | |
| D21089.1 | - | 9 | 1410 | 116 | ||
| EU530533.1 | - | 1 | 75 | 116 | ||
| HM113488.1 | - | 9 | 1309 | 116 | ||
| KJ534962.1 | - | 9 | 1305 | 116 | ||
| KJ535085.1 | - | 8 | 1181 | 116 | ||
| X65024.1 | - | 8 | 1145 | 116 | ||
More Information
Set Membership: |
|

XPC
Genomic Map