Assay Details
| Gene Symbol: ACTR3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017012460.1 | XP_016867949.1 | 10 - 11 | 1226 | 75 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ACTR3B| Gene Symbol: ACTR3B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017012460.1 | XP_016867949.1 | 10 - 11 | 1226 | 75 | |
Set Membership: |
|