Assay Details
| Gene Symbol: ZNF316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278559.1 | NP_001265488.1 | 9 | 3739 | 79 | |
| XM_006715630.3 | XP_006715693.1 | 8 | 3856 | 79 | ||
More Information
Set Membership: |
|

ZNF316
Genomic Map