Assay Details
| Gene Symbol: CENPN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001100624.2 | NP_001094094.2 | 11 | 112 | ||
| NM_001270473.1 | NP_001257402.1 | 10 | 112 | |||
| NM_001270474.1 | NP_001257403.1 | 10 | 112 | |||
| XM_006721236.3 | XP_006721299.1 | 11 | 112 | |||
| GenBank mRNA | AW085187.1 | - | 1 | 139 | 112 | |
More Information
Set Membership: |
|

CENPN
Genomic Map