Assay Details
| Gene Symbol: PRKAR1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_212471.2 | NP_997636.1 | 1 | 273 | 82 | |
| XM_011524985.2 | XP_011523287.1 | 1 | 275 | 82 | ||
| GenBank mRNA | BU167718.1 | - | 1 | 120 | 82 | |
| BU188258.1 | - | 1 | 73 | 82 | ||
| DA413808.1 | - | 1 | 273 | 82 | ||
| Y07642.1 | - | 1 | 53 | 82 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_212471.2, XM_011524985.2, Y07642.1 transcripts.
Set Membership: |
|

PRKAR1A
Genomic Map