Assay Details
| Gene Symbol: GFAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002055.4 | NP_002046.1 | 8 | 1270 | 60 | |
| XM_017024451.1 | XP_016879940.1 | 9 | 1341 | 60 | ||
| GenBank mRNA | AF419299.1 | - | 8 | 1209 | 60 | |
| AK098380.1 | - | 9 | 1156 | 60 | ||
| AK098758.1 | - | 6 | 909 | 60 | ||
| AK124465.1 | - | 1 | 131 | 60 | ||
| AK222683.1 | - | 8 | 1218 | 60 | ||
| AK296259.1 | - | 9 | 1167 | 60 | ||
| AK303728.1 | - | 9 | 1143 | 60 | ||
| AK315398.1 | - | 8 | 1224 | 60 | ||
| BC013596.1 | - | 8 | 1206 | 4185584 | 60 | |
| BC041765.1 | - | 8 | 1210 | 4186297 | 60 | |
| BC062609.1 | - | 8 | 1203 | 60 | ||
| BC127871.1 | - | 3 | 285 | 60 | ||
| J04569.1 | - | 8 | 1207 | 60 | ||
| M26638.1 | - | 3 | 152 | 60 | ||
| S40719.1 | - | 8 | 1218 | 60 | ||
More Information
Set Membership: |
|

GFAP
Genomic Map