Assay Details
| Gene Symbol: NR5A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011509381.2 | XP_011507683.1 | 1 | 1734 | 57 | |
| GenBank mRNA | AF124248.1 | - | 2 | 57 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011509381.2 transcripts.
Set Membership: |
|

NR5A2
Genomic Map