Assay Details
| Gene Symbol: FLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002016.1 | NP_002007.1 | 1 | 49 | 126 | |
| GenBank mRNA | L01090.1 | - | 1 | 50 | 126 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the L01090.1, NM_002016.1 transcripts.
Set Membership: |
|

FLG
Genomic Map