Assay Details
| Gene Symbol: IL17RA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001289905.1 | NP_001276834.1 | 7 | 843 | 74 | |
| NM_014339.6 | NP_055154.3 | 7 | 843 | 74 | ||
| GenBank mRNA | AF021799.1 | - | 1 | 115 | 74 | |
| BC011624.2 | - | 7 | 787 | 4053746 | 74 | |
| BX334547.2 | - | 7 | 846 | 74 | ||
| BX410574.2 | - | 7 | 759 | 74 | ||
| CT841520.1 | - | 7 | 766 | 74 | ||
| JF305973.1 | - | 7 | 710 | 74 | ||
| U58917.1 | - | 7 | 742 | 74 | ||
More Information
Set Membership: |
|

IL17RA
Genomic Map