Assay Details
| Gene Symbol: SOX11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003108.3 | NP_003099.1 | 1 | 36 | 53 | |
| GenBank mRNA | AB028641.1 | - | 1 | 36 | 53 | |
| BC025789.1 | - | 1 | 31 | 53 | ||
| U23752.1 | - | 1 | 56 | 53 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AB028641.1, BC025789.1, NM_003108.3, U23752.1 transcripts.
Set Membership: |
|

SOX11
Genomic Map